missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ETFDH Protein
A DNA sequence encoding the Homo sapiens (Human) ETFDH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7564-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001268666.1 | |
ETFDH Protein | |
Research Use Only | |
ETFDH | |
Recombinant Protein | |
Yeast | |
SSTSTVPRIT THYTIYPRDK DKRWEGVNME RFAEEADVVI VGAGPAGLSA AVRLKQLAVA HEKDIRVCLV EKAAQIGAHT LSGACLDPGA FKELFPDWKE KGAPLNTPVT EDRFGILTEK YRIPVPILPG LPMNNHGNYI VRLGHLVSWM GEQAEALGVE VYPGYAAAEV LFHDDGSVKG IATNDVGIQK DGAPKATFER GLELHAKVTI FAEGCHGHLA KQLYKKFDLR ANCEPQTYGI GLKELWVIDE KNWKPGRVDH TVGWPLDRHT YGGSFLYHLN EGEPLVALGL VVGLDYQNPY LSPFREFQRW KHHPSIRPTL EGGKRIAYGA RALNEGGFQS IPKLTFPGGL LIGCSPGFMN VPKIKGTHTA MKSGILAAES IFNQLTSENL QSKTIGLHVT EYEDNLKNSW VWKELYSVRN IRPSCHGVLG VYGGMIYTGI FYWILRGMEP WTLKHKGSDF ERLKPAKDCT PIEYPKPDGQ ISFDLLSSVA LSGTNHEHDQ PAHLTLRDDS IPVNRNLSIY DGPEQRFCPA GVYEFVPVEQ GDGFRLQINA QNCVHCKTCD IKDPSQNINW VVPEGGGGPA YNGM | |
Greater than 90% as determined by SDS-PAGE. |
2110 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |