missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Erythropoietin R Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 307-ER-050/CF
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human Erythropoietin R Protein is derived from NS0. The Recombinant Human Erythropoietin R Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
25 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
2057 | |
Erythropoietin R | |
Mouse myeloma cell line,NS0-derived human Erythropoietin R protein Ala25-Pro250 | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |