missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Erythropoietin / EPO Protein

Product Code. p-7148165
Click to view available options
Quantity:
10 μg
100 μg
1500 IU
2 μg
2000 IU
250 μg
37500 IU
Unit Size:
100µg
10µg
1500 units
2000 units
250µg
2µg
37500 units
This item is not returnable. View return policy

Product Code. 15973578

Brand: enQuireBio™ QP10347ma2ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human Erythropoietin / EPO, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000790.2
Gene ID (Entrez) 2056
Name Erythropoietin / EPO Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol EPO
Protein Length Partial (see sequence information for more details).
Biological Activity The ED50 determined by a cell proliferation assay using human megakaryoblastic leukemia cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species Mammal
Protein Tag FC
Buffer 0.2um filtered sodium citrate buffer (1 liter of ddH2O containing 5.9 g of sodium citrate, 5.8 g of sodium chloride and 0.06 g of citric acid), lyophilized
Purity or Quality Grade Greater than 98% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.