missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Erythropoietin / EPO Protein

Product Code. 15923588
Click to view available options
Quantity:
1500 IU
2000 IU
37500 IU
500 IU
Unit Size:
1500 units
2000 units
37500 units
500 units
This item is not returnable. View return policy

Product Code. 15923588

Brand: enQuireBio™ QP10349ma500IU

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human Erythropoietin / EPO, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000790.2
Gene ID (Entrez) 2056
Name Erythropoietin / EPO Protein
Quantity 500 IU
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol EPO
Protein Length Partial (see sequence information for more details).
Biological Activity The Specific Activity was measured by the stimulation of reticulocyte production in normocyth-aemic mice and was found to be no less than 1.5 M 105 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species Mammal
Protein Tag Untagged
Sequence APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR
Buffer 0.2um filtered sodium citrate buffer (1 liter of ddH2O containing 5.9 g of sodium citrate, 5.8 g of sodium chloride and 0.06 g of citric acid), lyophilized
Purity or Quality Grade Greater than 98% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.