missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Erythropoietin / EPO Protein
Click to view available options
Quantity:
1500 IU
2000 IU
37500 IU
500 IU
Unit Size:
1500 units
2000 units
37500 units
500 units
Specifications
Specifications
| Accession Number | NP_000790.2 |
| Gene ID (Entrez) | 2056 |
| Name | Erythropoietin / EPO Protein |
| Quantity | 2000 IU |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
| Gene Symbol | EPO |
| Protein Length | Partial (see sequence information for more details). |
| Biological Activity | The Specific Activity was measured by the stimulation of reticulocyte production in normocyth-aemic mice and was found to be no less than 1.5 M 105 IU/mg. |
| Product Type | Recombinant Protein |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction