missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ER membrane protein complex subunit 9 Protein

A DNA sequence encoding the Homo sapiens (Human) ER membrane protein complex subunit 9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8297-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16073851

  • £384.00 / 100µg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057133.2
ER membrane protein complex subunit 9 Protein
Research Use Only
EMC9
Recombinant Protein
E. coli
MGEVEISALA YVKMCLHAAR YPHAAVNGLF LAPAPRSGEC LCLTDCVPLF HSHLALSVML EVALNQVDVW GAQAGLVVAG YYHANAAVND QSPGPLALKI AGRIAEFFPD AVLIMLDNQK LVPQPRVPPV IVLENQGLRW VPKDKNLVMW RDWEESRQMV GALLEDRAHQ HLVDFDCHLD DIRQDWTNQR LNTQITQWVG PTNGNGNA
Greater than 90% as determined by SDS-PAGE.
51016
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers