missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EPHA2 Protein

A cDNA sequence encoding the EPHA2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11796-HIS-HEK-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988398

  • £99.15 / 2µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EPHA2 Protein
Research Use Only
Recombinant Protein
Human (HEK293)
AQGKEVVLLD FAAAGGELGW LTHPYGKGWD LMQNIMNDMP IYMYSVCNVM SGDQDNWLRT NWVYRGEAER IFIELKFTVR DCNSFPGGAS SCKETFNLYY AESDLDYGTN FQKRLFTKID TIAPDEITVS SDFEARHVKL NVEERSVGPL TRKGFYLAFQ DIGACVALLS VRVYYKKCPE LLQGLAHFPE TIAGSDAPSL ATVAGTCVDH AVVPPGGEEP RMHCAVDGEW LVPIGQCLCQ AGYEKVEDAC QACSPGFFKF EASESPCLEC PEHTLPSPEG ATSCECEEGF FRAPQDPASM PCTRPPSAPH YLTAVGMGAK VELRWTPPQD SGGREDIVYS VTCEQCWPES GECGPCEASV RYSEPPHGLT RTSVTVSDLE PHMNYTFTVE ARNGVSGLVT SRSFRTASVS INQTEPPKVR LEGRSTTSLS VSWSIPPPQQ SRVWKYEVTY RKKGDSNSYN VRRTEGFSVT LDDLAPDTTY LVQVQALTQE GQGAGSKVHE FQTLSPEKLH HHHHH
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
EPHA2 was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline and 5% (w/v) trehalose.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers