missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human EpCAM/TROP1 Fc Alexa Fluor™ 488 Protein

Product Code. 18756746
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy

Product Code. 18756746

Brand: R&D Systems™ AFG960020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by flow cytometry for its ability to bind Human EpCAM/TROP-1 Antibody conjugated fluorescent beads at 0.500-2.00 μg/mL (100 μL/well). Please Note: Optimal dilutions should be determined by each laboratory for each application.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CAA32870
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 4072
Molecular Weight (g/mol) M.W. Observed: 60-70 kDa, under reducing conditions., M.W. Theoretical: 54 kDa
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human EpCAM/TROP1 protein Human EpCAM (Gln24-Lys265) (N-terminus) Accession # CAA32870.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Gene Alias 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Conjugate Alexa Fluor 488
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EpCAM/TROP1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.