missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ENPP-7/Alk-SMase Protein

Product Code. 18106200
Click to view available options
:
10ug; Carrier Free
This item is not returnable. View return policy

Product Code. 18106200

Brand: R&D Systems™ 4924EN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze sphingomyelin to ceramide and phosphorylcholine. The phosphorylcholine is cleaved by Recombinant Human Alkaline Phosphatase/ALPL (Catalog # 2909-AP ) and the phosphate is detected/measured by a Malachite Green Phosphate Detection Kit (Catalog # DY996 ). The specific activity is >14,000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_848638
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 339221
Molecular Weight (g/mol) M.W. (Observed): 60-70 kDa, reducing conditions; M.W. (theoretical): 49 kDa
Name ENPP-7/Alk-SMase
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human ENPP-7/Alk-SMase protein Ala22-Ser439, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol ENPP7
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein ENPP-7/Alk-SMase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.