missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Endostatin protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting the FGF basic-dependent proliferation of HUVEC migration is less than 2.0 μg/mL in the presence of Anti-Human Endostatin Polyclonal Antibody.
Specifications
Specifications
| Accession Number | P39059 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 80781 |
| Molecular Weight (g/mol) | 20.1 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of Endostatin as determined by LAL method. |
| Gene Alias | antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?