missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human EMMPRIN/CD147 (aa 138-323) His Protein, CF

Product Code. 18320528
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320528 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320528 Supplier R&D Systems™ Supplier No. 10611EM050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isoform 1, HEK293 Expressed

Measured by the ability of the immobilized protein to induce active MMP-1 secretion by NHLF human normal lung fibroblasts. The ED50 for this effect is 0.70-5.60 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35613.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 682
Molecular Weight (g/mol) M.W. (Observed): 25-41 kDa, under reducing conditions, M.W. (Theoretical): 21 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human EMMPRIN/CD147 protein Glu138-Ala323, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EMMPRIN/CD147
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.