missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fully biologically active when compared to standard. The ED50 as determined by the apoptotic effect using serum free human MCF-7 cells is less than 40 ng/mL, corresponding to a specific activity of >2.5 x 10^4 IU/mg.
Specifications
Specifications
| Accession Number | Q12904, Q12904, Q12904 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 9255 |
| Molecular Weight (g/mol) | 18.2 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of EMAP-II/AIMP1 as determined by LAL method. |
| Gene Alias | aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?