missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Elongation factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Elongation factor 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5962-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001952.1 | |
Elongation factor 2 Protein | |
Research Use Only | |
EEF2 | |
Recombinant Protein | |
E. coli | |
VNFTVDQIRA IMDKKANIRN MSVIAHVDHG KSTLTDSLVC KAGIIASARA GETRFTDTRK DEQERCITIK STAISLFYEL SENDLNFIKQ SKDGAGFLIN LIDSPGHVDF SSEVTAALRV TDGALVVVDC VSGVCVQTET VLRQAIAERI KPVLMMNKMD RALLELQLEP EELYQTFQRI VENVNVIIST YGEGESGPMG NIMIDPVLGT VGFGSGLHGW AFTLKQFAEM YVAKFAAKGE GQLGPAERAK KVEDMMKKLW GDRYFDPANG KFSKSATSPE GKKLPRTFCQ LILDPIFKVF DAIMNFKKEE TAKLIEKLDI KLDSEDKDKE GKPLLKAVMR RWLPAGDALL QMITIHLPSP VTAQKYRCEL LYEGPPDDEA AMGIKSCDPK GPLMMYISKM VPTSDKGRFY AFGRVFSGLV STGLKVRIMG PNYTPGKKED LYLKPIQRTI LMMGRYVEPI EDVPCGNIVG LVGVDQFLVK TGTITTFEHA HNMRVMKFSV SPVVRVAVEA KNPADLPKLV EGLKRLAKSD PMVQCIIEES GEHIIAGAGE LHLEICLKDL EEDHACIPIK KSDPVVSYRE TVSEESNVLC LSKSPNKHNR LYMKARPFPD GLAEDIDKGE VSARQELKQR ARYLAEKYEW DVAEARKIWC FGPDGTGPNI LTDITKGVQY LNEIKDSVVA GFQWATKEGA LCEENMRGVR FDVHDVTLHA DAIHRGGGQI IPTARRCLYA SVLTAQPRLM EPIYLVEIQC PEQVVGGIYG VLNRKRGHVF EESQVAGTPM FVVKAYLPVN ESFGFTADLR SNTGGQAFPQ CVFDHWQILP GDPFDNSSRP SQVVAETRKR KGLKEGIPAL DNFLDKL | |
Greater than 90% as determined by SDS-PAGE. |
1938 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |