missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Elongation factor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Elongation factor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5962-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968959

  • £380.58 / 100µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001952.1
Elongation factor 2 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
1938
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EEF2
Recombinant Protein
E. coli
VNFTVDQIRA IMDKKANIRN MSVIAHVDHG KSTLTDSLVC KAGIIASARA GETRFTDTRK DEQERCITIK STAISLFYEL SENDLNFIKQ SKDGAGFLIN LIDSPGHVDF SSEVTAALRV TDGALVVVDC VSGVCVQTET VLRQAIAERI KPVLMMNKMD RALLELQLEP EELYQTFQRI VENVNVIIST YGEGESGPMG NIMIDPVLGT VGFGSGLHGW AFTLKQFAEM YVAKFAAKGE GQLGPAERAK KVEDMMKKLW GDRYFDPANG KFSKSATSPE GKKLPRTFCQ LILDPIFKVF DAIMNFKKEE TAKLIEKLDI KLDSEDKDKE GKPLLKAVMR RWLPAGDALL QMITIHLPSP VTAQKYRCEL LYEGPPDDEA AMGIKSCDPK GPLMMYISKM VPTSDKGRFY AFGRVFSGLV STGLKVRIMG PNYTPGKKED LYLKPIQRTI LMMGRYVEPI EDVPCGNIVG LVGVDQFLVK TGTITTFEHA HNMRVMKFSV SPVVRVAVEA KNPADLPKLV EGLKRLAKSD PMVQCIIEES GEHIIAGAGE LHLEICLKDL EEDHACIPIK KSDPVVSYRE TVSEESNVLC LSKSPNKHNR LYMKARPFPD GLAEDIDKGE VSARQELKQR ARYLAEKYEW DVAEARKIWC FGPDGTGPNI LTDITKGVQY LNEIKDSVVA GFQWATKEGA LCEENMRGVR FDVHDVTLHA DAIHRGGGQI IPTARRCLYA SVLTAQPRLM EPIYLVEIQC PEQVVGGIYG VLNRKRGHVF EESQVAGTPM FVVKAYLPVN ESFGFTADLR SNTGGQAFPQ CVFDHWQILP GDPFDNSSRP SQVVAETRKR KGLKEGIPAL DNFLDKL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers