missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF5A Protein
A cDNA sequence encoding the EIF5A was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | EIF5A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948328
|
enQuireBio™
QP11775-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938328
|
enQuireBio™
QP11775-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928328
|
enQuireBio™
QP11775-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
EIF5A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The EIF5A protein solution (1 mg/ml) contains 50mM Tris-HCl, pH 7.5 and 10% Glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MADDLDFETG DAGASATFPM QCSALRKNGF VVLKGRPCKI VEMSTSKTGK HGHAKVHLVG IDIFTGKKYE DICPSTHNMD VPNIKRNDFQ LIGIQDGYLS LLQDSGEVRE DLRLPEGDLG KEIEQKYDCG EEILITVLSA MTEEAAVAIK AMAK | |
Greater than 95.0% as determined by SDS-PAGE. |