missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF1AX Protein
A cDNA sequence encoding the EIF1AX was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11763-1mg
Additional Details : Weight : 0.01000kg
Specifications
EIF1AX Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPKNKGKGGK NRRRGKNENE SEKRELVFKE DGQEYAQVIK MLGNGRLEAM CFDGVKRLCH IRGKLRKKVW INTSDIILVG LRDYQDNKAD VILKYNADEA RSLKAYGELP EHAKINETDT FGPGDDDEIQ FDDIGDDDED IDDI | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EIF1AX protein solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0) containing 5mM DTT, 200mM NaCl and 40% glycerol. |