missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EHF Protein
A cDNA sequence encoding the EHF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11762-10ug
Additional Details : Weight : 0.01000kg
Specifications
EHF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMILEGGG VMNLNPGNNL LHQPPAWTDS YSTCNVSSGF FGGQWHEIHP QYWTKYQVWE WLQHLLDTNQ LDANCIPFQE FDINGEHLCS MSLQEFTRAA GTAGQLLYSN LQHLKWNGQC SSDLFQSTHN VIVKTEQTEP SIMNTWKDEN YLYDTNYGST VDLLDSKTFC RAQISMTTTS HLPVAESPDM KKEQDPPAKC HTKKHNPRGT HLWEFIRDIL LNPDKNPGLI KWEDRSEGVF RFLKSEAVAQ LWGKKKNNSS MTYEKLSRAM RYYYKREILE RVDGRRLVYK FGKNARGWRE NEN | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EHF solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |