missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Egl nine homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Egl nine homolog 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5968-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_444274.1 | |
Egl nine homolog 2 Protein | |
Research Use Only | |
EGLN2 | |
Recombinant Protein | |
E. coli | |
MVACYPGNGL GYVRHVDNPH GDGRCITCIY YLNQNWDVKV HGGLLQIFPE GRPVVANIEP LFDRLLIFWS DRRNPHEVKP AYATRYAITV WYFDAKERAA AKDKYQLASG QKGVQVPVSQ PPTPT | |
Greater than 90% as determined by SDS-PAGE. |
112398 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |