missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human EGFR Fc Chimera Alexa Fluor™ 647 Protein

Product Code. 18739956
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy

Product Code. 18739956

Brand: R&D Systems™ AFR344020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by flow cytometry for its ability to bind anti-human EGFR Monoclonal Antibody conjugated beads. The concentration of Recombinant Human EGFR Fc-tag Alexa Fluor™ 647 (Catalog # AFR344) that produces 50% of the binding response is 0.750-15.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CAA25240.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1956
Molecular Weight (g/mol) M.W.-observed: 118-134 kDa, under reducing conditions., M.W.-theroretical: 95 kDa (monomer)
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human EGFR protein Human EGFR (Leu25-Ser645) (C-terminus) Accession # CAA25240.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Conjugate Alexa Fluor 647
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EGFR
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.