missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human eEF1A1 Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Quantity:
0.1 mg
0.5 mg
Packungsgröße:
0.1mg
0.5mg
Beschreibung
An un-tagged denatured recombinant protein corresponding to the amino acids 1-462 of Human eEF1A1 The Recombinant Human eEF1A1 Protein is derived from E. coli. The Recombinant Human eEF1A1 Protein has been validated for the following applications: SDS-Page.
Spezifikation
Spezifikation
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.4M UREA, 10% glycerol |
| Gene ID (Entrez) | 1915 |
| Molecular Weight (g/mol) | TMW: 50kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | CCS3, CCS-3, cervical cancer suppressor 3, CTCL tumor antigen, EE1A1, EEF-1, EEF1A, eEF1A-1, EF1AHNGC:16303, EF1a-like protein, EF-1-alpha-1, EF-Tu, elongation factor 1 alpha subunit, elongation factor 1-alpha 1, Elongation factor Tu, Eukaryotic elongation factor 1 A-1, eukaryotic translation elongation factor 1 alpha 1, eukaryotic translation elongation factor 1 alpha 1-like 14, FLJ25721, glucocorticoid receptor AF-1 specific elongation factor, GRAF-1EF, LENG7, leukocyte receptor cluster (LRC) member 7, Leukocyte receptor cluster member 7, MGC102687, MGC131894, MGC16224, prostate tumor-inducing protein 1, PTI1, translation elongation factor 1 alpha 1-like 14 |
| Common Name | eEF1A1 |
| Conjugate | Unconjugated |
| Mehr anzeigen |
For research use only and is not approved for use in humans or in clinical diagnosis
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur