missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EDN2 Protein

A cDNA sequence encoding the EDN2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11746-HIS-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968228

  • £99.15 / 5µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EDN2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQAAATLE QPASSSHAQG THLRLRRCSC SSWLDKECVY FCHLDIIWVN TPEQTAPYGL GNPPRRRRRS LPRRCQCSSA RDPACATFCL RRPWTEAGAV PSRKSPADVF QTGKTGATTG ELLQRLRDIS TVKSLFAKRQ QEAMREPRST HSRWRKR
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
EDN2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers