missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ECHS1 Protein
A cDNA sequence encoding the ECHS1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11739-20ug
Additional Details : Weight : 0.01000kg
Specifications
1892 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASGANFEYI IAEKRGKNNT VGLIQLNRPK ALNALCDGLI DELNQALKIF EEDPAVGAIV LTGGDKAFAA GADIKEMQNL SFQDCYSSKF LKHWDHLTQV KKPVIAAVNG YAFGGGCELA MMCDIIYAGE KAQFAQPEIL IGTIPGAGGT QRLTRAVGKS LAMEMVLTGD RISAQDAKQA GLVSKICPVE TLVEEAIQCA EKIASNSKIV VAMAKESVNA AFEMTLTEGS KLEKKLFYST FATDDRKEGM TAFVEKRKAN FKDQ | |
Greater than 95.0% as determined by SDS-PAGE. |
ECHS1 Protein | |
Research Use Only | |
ECHS1 | |
Human | |
His | |
The ECHS1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8), 1mM DTT,0.1M NaCl and 20% glycerol. |