missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ECHDC1 Protein
A cDNA sequence encoding the ECHDC1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11738-5ug
Additional Details : Weight : 0.01000kg
Specifications
55862 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MALKQEMAKS LLKTASLSGR TKLLHQTGLS LYSTSHGFYE EEVKKTLQQF PGGSIDLQKE DNGIGILTLN NPSRMNAFSG VMMLQLLEKV IELENWTEGK GLIVRGAKNT FSSGSDLNAV KSLGTPEDGM AVCMFMQNTL TRFMRLPLIS VALVQGWALGGGAEFTTACD FRLMTPESKI RFVHKEMGII PSWGGTTRLV EIIGSRQALK VLSGALKLDS KNALNIGMVE EVLQSSDETK SLEEAQEWLK QFIQGPPEVI RALKKSVCSG RELYLEEALQ NERDLLGTVW GGPANLEAIA KKGKFNK | |
Greater than 95% as determined by SDS-PAGE. |
ECHDC1 Protein | |
Research Use Only | |
ECHDC1 | |
Human | |
His | |
The ECHDC1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1M NaCl. |