missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase SMURF2 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase SMURF2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8114-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_073576.1 | |
E3 ubiquitin-protein ligase SMURF2 Protein | |
Research Use Only | |
SMURF2 | |
Recombinant Protein | |
E. coli | |
MSNPGGRRNG PVKLRLTVLC AKNLVKKDFF RLPDPFAKVV VDGSGQCHST DTVKNTLDPK WNQHYDLYIG KSDSVTISVW NHKKIHKKQG AGFLGCVRLL SNAINRLKDT GYQRLDLCKL GPNDNDTVRG QIVVSLQSRD RIGTGGQVVD CSRLFDNDLP DGWEERRTAS GRIQYLNHIT RTTQWERPTR PASEYSSPGR PLSCFVDENT PISGTNGATC GQSSDPRLAE RRVRSQRHRN YMSRTHLHTP PDLPEGYEQR TTQQGQVYFL HTQTGVSTWH DPRVPRDLSN INCEELGPLP PGWEIRNTAT GRVYFVDHNN RTTQFTDPRL SANLHLVLNR QNQLKDQQQQ QVVSLCPDDT ECLTVPRYKR DLVQKLKILR QELSQQQPQA GHCRIEVSRE EIFEESYRQV MKMRPKDLWK RLMIKFRGEE GLDYGGVARE WLYLLSHEML NPYYGLFQYS RDDIYTLQIN PDSAVNPEHL SYFHFVGRIM GMAVFHGHYI DGGFTLPFYK QLLGKSITLD DMELVDPDLH NSLVWILEND ITGVLDHTFC VEHNAYGEII QHELKPNGKS IPVNEENKKE YVRLYVNWRF LRGIEAQFLA LQKGFNEVIP QHLLKTFDEK ELELIICGLG KIDVNDWKVN TRLKHCTPDS NIVKWFWKAV EFFDEERRAR LLQFVTGSSR VPLQGFKALQ GAAGPRLFTI HQIDACTNNL PKAHTCFNRI DIPPYESYEK LYEKLLTAIE ETCGFAVE | |
Greater than 90% as determined by SDS-PAGE. |
64750 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |