missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RAD18 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase RAD18, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_064550.3 |
---|---|
Gene ID (Entrez) | 56852 |
Name | E3 ubiquitin-protein ligase RAD18 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063381
|
enQuireBio™
QP8224-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083381
|
enQuireBio™
QP8224-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073381
|
enQuireBio™
QP8224-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093381
|
enQuireBio™
QP8224-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003391
|
enQuireBio™
QP8224-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013391
|
enQuireBio™
QP8224-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_064550.3 | |
E3 ubiquitin-protein ligase RAD18 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
56852 | |
Research Use Only | |
RAD18 | |
Recombinant Protein | |
E. coli | |
MDSLAESRWP PGLAVMKTID DLLRCGICFE YFNIAMIIPQ CSHNYCSLCI RKFLSYKTQC PTCCVTVTEP DLKNNRILDE LVKSLNFARN HLLQFALESP AKSPASSSSK NLAVKVYTPV ASRQSLKQGS RLMDNFLIRE MSGSTSELLI KENKSKFSPQ KEASPAAKTK ETRSVEEIAP DPSEAKRPEP PSTSTLKQVT KVDCPVCGVN IPESHINKHL DSCLSREEKK ESLRSSVHKR KPLPKTVYNL LSDRDLKKKL KEHGLSIQGN KQQLIKRHQE FVHMYNAQCD ALHPKSAAEI VREIENIEKT RMRLEASKLN ESVMVFTKDQ TEKEIDEIHS KYRKKHKSEF QLLVDQARKG YKKIAGMSQK TVTITKEDES TEKLSSVCMG QEDNMTSVTN HFSQSKLDSP EELEPDREED SSSCIDIQEV LSSSESDSCN SSSSDIIRDL LEEEEAWEAS HKNDLQDTEI SPRQNRRTRA AESAEIEPRN KRNRN | |
Greater than 90% as determined by SDS-PAGE. |