missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DYNLT1 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | DYNLT1 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MEDYQAAEET AFVVDEVSNI VKEAIESAIG GNAYQHSKVN QWTTNVVEQT LSQLTKLGKP FKYIVTCVIM QKNGAGLHTA SSCFWDSSTD GSCTVRWENK TMYCIVSAFG LSI |
| Buffer | DYNLT1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction