missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dynamin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Dynamin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5939-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968819

  • £1265.00 / 500µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001005336.1
Dynamin-1 Protein
Research Use Only
DNM1
Recombinant Protein
Yeast
GNRGMEDLIP LVNRLQDAFS AIGQNADLDL PQIAVVGGQS AGKSSVLENF VGRDFLPRGS GIVTRRPLVL QLVNATTEYA EFLHCKGKKF TDFEEVRLEI EAETDRVTGT NKGISPVPIN LRVYSPHVLN LTLVDLPGMT KVPVGDQPPD IEFQIRDMLM QFVTKENCLI LAVSPANSDL ANSDALKVAK EVDPQGQRTI GVITKLDLMD EGTDARDVLE NKLLPLRRGY IGVVNRSQKD IDGK
Greater than 90% as determined by SDS-PAGE.
1759
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers