missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynactin subunit 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5919-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006562.1 | |
Dynactin subunit 6 Protein | |
Research Use Only | |
DCTN6 | |
Recombinant Protein | |
E. coli | |
MAEKTQKSVK IAPGAVVCVE SEIRGDVTIG PRTVIHPKAR IIAEAGPIVI GEGNLIEEQA LIINAYPDNI TPDTEDPEPK PMIIGTNNVF EVGCYSQAMK MGDNNVIESK AYVGRNVILT SGCIIGACCN LNTFEVIPEN TVIYGADCLR RVQTERPQPQ TLQLDFLMKI LPNYHHLKKT MKGSSTPVKN | |
Greater than 80% as determined by SDS-PAGE. |
10671 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |