missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dynactin subunit 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9937-ye-100ug

 View more versions of this product

Product Code. 16049041

  • £467.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001129115.1
Dynactin subunit 4 Protein
Research Use Only
DCTN4
Recombinant Protein
Yeast
ASLLQSDRVL YLVQGEKKVR APLSQLYFCR YCSELRSLEC VSHEVDSHYC PSCLENMPSA EAKLKKNRCA NCFDCPGCMH TLSTRATSIS TQLPDDPAKT TMKKAYYLAC GFCRWTSRDV GMADKSVASG GWQEPENPHT QRMNKLIEYY QQLAQKEKVE RDRKKLARRR NYMPLAFSDK YGLGTRLQRP RAGASISTLA GLSLKEGEDQ KEIKIEPAQA VDEVEPLPED YYTRPVNLTE VTTLQQRLLQ PDFQPVCASQ LYPRHKHLLI KRSLRCRKCE HNLSKPEFNP TSIKFKIQLV AVNYIPEVRI MSIPNLRYMK ESQVLLTLTN PVENLTHVTL FECEEGDPDD INSTAKVVVP PKELVLAGKD AAAEYDELAE PQDFQDDPDI IAFRKANKVG IFIKVTPQRE EGEVTVCFKM KHDFKNLAAP IRPIEESDQG TEVIWLTQHV ELSLGPLLP
Greater than 90% as determined by SDS-PAGE.
51164
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers