missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dual specificity protein phosphatase 26 Protein

A DNA sequence encoding the Homo sapiens (Human) Dual specificity protein phosphatase 26, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8061-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082431

  • £384.00 / 100µg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001292044.1
Dual specificity protein phosphatase 26 Protein
Research Use Only
DUSP26
Recombinant Protein
E. coli
MCPGNWLWAS MTFMARFSRS SSRSPVRTRG TLEEMPTVQH PFLNVFELER LLYTGKTACN HADEVWPGLY LGDQDMANNR RELRRLGITH VLNASHSRWR GTPEAYEGLG IRYLGVEAHD SPAFDMSIHF QTAADFIHRA LSQPGGKILV HCAVGVSRSA TLVLAYLMLY HHLTLVEAIK KVKDHRGIIP NRGFLRQLLA LDRRLRQGLE A
Greater than 90% as determined by SDS-PAGE.
78986
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers