missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dual specificity protein phosphatase 22 Protein
A DNA sequence encoding the Homo sapiens (Human) Dual specificity protein phosphatase 22, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8074-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273484.1 | |
Dual specificity protein phosphatase 22 Protein | |
Research Use Only | |
DUSP22 | |
Recombinant Protein | |
E. coli | |
GNGMNKILPG LYIGNFKDAR DAEQLSKNKV THILSVHDSA RPMLEGVKYL CIPAADSPSQ NLTRHFKESI KFIHECRLRG ESCLVHCLAG VSRSVTLVIA YIMTVTDFGW EDALHTVRAG RSCANPNVGF QRQLQEFEKH EVHQYRQWLK EEYGESPLQD AEEAKNILAA PGILKFWAFL RRL | |
Greater than 80% as determined by SDS-PAGE. |
56940 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |