missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DPPA3 Protein
A cDNA sequence encoding the DPPA3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11698-2ug
Additional Details : Weight : 0.01000kg
Specifications
DPPA3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDPSQFN PTYIPGSPQM LTEENSRDDS GASQISSETL IKNLSNLTIN ASSESVSPLS EALLRRESVG AAVLREIEDE WLYSRRGVRT LLSVQREKMA RLRYMLLGGV RTHERRPTNK EPKGVKKESR PFKCPCSFCV SNGWDPSENA RIGNQDTKPL QP | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DPPA3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |