missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DOK4 Protein
A cDNA sequence encoding the DOK4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11695-5ug
Additional Details : Weight : 0.01000kg
Specifications
DOK4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMATNFSD IVKQGYVKMK SRKLGIYRRC WLVFRKSSSK GPQRLEKYPD EKSVCLRGCP KVTEISNVKC VTRLPKETKR QAVAIIFTDD SARTFTCDSE LEAEEWYKTL SVECLGSRLN DISLGEPDLL APGVQCEQTD RFNVFLLPCP NLDVYGECKL QITHENIYLW DIHNPRVKLV SWPLCSLRRY GRDATRFTFE AGRMCDAGEG LYTFQTQEGE QIYQRVHSAT LAIAEQHKRV LLEMEKNVRL LNKGTEHYSY PCTPTTMLPR SAYWHHITGS QNIAEASSYA GEGYGAAQAS SETDLLNRFI LLKPKPSQGD SSEAKTPSQ | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DOK4 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |