missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Docking protein 1

A DNA sequence encoding the Homo sapiens (Human) Docking protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5943-ec-1mg

 View more versions of this product

Product Code. 15988839

  • £1735.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_060901.2
Docking protein 1
Research Use Only
DOK5
Recombinant Protein
E. coli
MECVGTRIND ISLGEPDLLA TGVEREQSER FNVYLMPSPN LDVHGECALQ ITYEYICLWD VQNPRVKLIS WPLSALRRYG RDTTWFTFEA GRMCETGEGL FIFQTRDGEA IYQKVHSAAL AIAEQHERLL QSVKNSMLQM KMSERAASLS TMVPLPRSAY WQHITRQHST GQLYRLQDVS SPLKLHRTET FPAYRSEH
Greater than 90% as determined by SDS-PAGE.
55816
1 mg
Not determined.
Partial (see sequence information for more details). of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers