missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNAL1 Protein
A cDNA sequence encoding the DNAL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11689-1mg
Additional Details : Weight : 0.01000kg
Specifications
DNAL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAKATTI KEALARWEEK TGQRPSEAKE IKLYAQIPPI EKMDASLSML ANCEKLSLST NCIEKIANLN GLKNLRILSL GRNNIKNLNG LEAVGDTLEE LWISYNFIEK LKGIHIMKKL KILYMSNNLV KDWAEFVKLA ELPCLEDLVF VGNPLEEKHS AENNWIEEAT KRVPKLKKLD GTPVIKGDEE EDN | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DNAL1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |