missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA replication licensing factor MCM2 Protein

Product Code. 16062660
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16062660

Brand: enQuireBio™ QP6349ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) DNA replication licensing factor MCM2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_004517.2
Gene ID (Entrez) 4171
Name DNA replication licensing factor MCM2 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol MCM2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence AESSESFTMA SSPAQRRRGN DPLTSSPGRS SRRTDALTSS PGRDLPPFED ESEGLLGTEG PLEEEEDGEE LIGDGMERDY RAIPELDAYE AEGLALDDED VEELTASQRE AAERAMRQRD REAGRGLGRM RRGLLYDSDE EDEERPARKR RQVERATEDG EEDEEMIESI ENLEDLKGHS VREWVSMAGP RLEIHHRFKN FLRTHVDSHG HNVFKERISD MCKENRESLV VNYEDLAARE HVLAYFLPEA PAELLQIFDE AALEVVLAMY PKYDRITNHI HVRISHLPLV EELRSLRQLH LNQLIRTSGV VTSCTGVLPQ LSMVKYNCNK CNFVLGPFCQ SQNQEVKPGS CPECQSAGPF EVNMEETIYQ NYQRIRIQES PGKVAAGRLP RSKDAILLAD LVDSCKPGDE IELTGIYHNN YDGSLNTANG FPVFATVILA NHVAKKDNKV AVGELTDEDV KMITSLSKDQ QIGEKIFASI APSIYGHEDI KRGLALALFG GEPKNPGGKH KVRGDINVLL CGDPGTAKSQ FLKYIEKVSS RAIFTTGQGA SAVGLTAYVQ RHPVSREWTL EAGALVLADR GVCLIDEFDK MNDQDRTSIH EAMEQQSISI SKAGIVTSLQ ARCTVIAAAN PIGGRYDPSL TFSENVDLTE PIISRFDILC VVRDTVDPVQ DEMLARFVVG SHVRHHPSNK EEEGLANGSA AEPAMPNTYG VEPLPQEVLK KYIIYAKERV HPKLNQMDQD KVAKMYSDLR KESMATGSIP ITVRHIESMI RMAEAHARIH LRDYVIEDDV NMAIRVMLES FIDTQKFSVM RSMRKTFARY LSFRRDNNEL LLFILKQLVA EQVTYQRNRF GAQQDTIEVP EKDLVDKARQ INIHNLSAFY DSELFRMNKF SHDLKRKMIL QQF
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.