missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human DNA Polymerase gamma GST (N-Term) Protein
Shop All Bio Techne ProductsDescription
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 5428 |
| Molecular Weight (g/mol) | 37.73 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE |
| Gene Symbol | POLG |
| Research Category | DNA Polymerases, DNA Repair |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?