missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human DLL4 Fc Chimera Biotinylated Protein, CF

Product Code. 30485267
Click to view available options
:
50 μg
This item is not returnable. View return policy

Product Code. 30485267

Brand: R&D Systems™ BT10185050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human DLL4 Fc Chimera (Catalog # BT10185) binds to Recombinant Human Notch-1 Fc Chimera (Catalog # 3647-TK ) with an ED50 of 7.00-84.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 54567
Molecular Weight (g/mol) Observed: 83-98 kDa, under reducing conditions.,Theoretical: 81 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human DLL4 protein Human DLL4 (Ser27-Pro524) (N-terminus) Accession # Q9NR61 IEGRMDP Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4
Conjugate Biotin
Reconstitution Reconstitute at 250 μg/mL in water.
Protein DLL4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.