missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DKK3 Protein
A cDNA sequence encoding the DKK3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11668-HIS-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
DKK3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHYHH SSGLVPRGSH MGSMAPAPTA TSAPVKPGPA LSYPQEEATL NEMFREVEEL MEDTQHKLRS AVEEMEAEEA AAKASSEVNL ANLPPSYHNE TNTDTKVGNN TIHVHREIHK ITNNQTGQMV FSETVITSVG DEEGRRSHEC IIDEDCGPSM YCQFASFQYT CQPCRGQRML CTRDSECCGD QLCVWGHCTK MATRGSNGTI CDNQRDCQPG LCCAFQRGLL FPVCTPLPVE GELCHDPASR LLDLITWELE PDGALDRCPC ASGLLCQPHS HSLVYVCKPT FVGSRDQDGE ILLPREVPDE YEVGSFMEEV RQELEDLERS LTEEMALREP AAAAAALLGG EEI | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DKK3 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1M Urea, 1mM DTT and 10% glycerol. |