missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Dkk-2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 6628-DK-010

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18116210

  • £278.61 / 10µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Dkk-2 Protein is derived from CHO. The Recombinant Human Dkk-2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Bioactivity
27123
Dkk-2
Chinese Hamster Ovary cell line,CHO-derived human Dkk-2 protein Met1-Ile259
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
25.8 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers