missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Dkk-1 Protein, CF

Product Code. 18796165
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
50 μg
Unit Size:
100µg
1mg
20µg
250µg
50µg
This item is not returnable. View return policy

Product Code. 18796165

Brand: R&D Systems™ 11387DK01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its ability to inhibit Wnt induced TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 8.00-96.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O94907.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 22943
Molecular Weight (g/mol) M.W. (Observed): 38-42 kDa, under reducing conditions., M.W. (Theroretical): 26 kDa
Quantity 1 mg
Source Human embryonic kidney cell, HEK293-derived human Dkk-1 protein Thr32-His266
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like
Conjugate Unconjugated
Reconstitution Reconstitute the 20 μg size at 200 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Dkk-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.