missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Dkk-1 Protein, CF

Product Code. 18785755
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
50 μg
Unit Size:
100µg
1mg
20µg
250µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18785755 100 μg 100µg
18796165 1 mg 1mg
18796055 20 μg 20µg
18796105 50 μg 50µg
18796005 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18785755 Supplier R&D Systems™ Supplier No. 11387DK100
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Measured by its ability to inhibit Wnt induced TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 8.00-96.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O94907.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 22943
Molecular Weight (g/mol) M.W. (Observed): 38-42 kDa, under reducing conditions., M.W. (Theroretical): 26 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human Dkk-1 protein Thr32-His266
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like
Conjugate Unconjugated
Reconstitution Reconstitute the 20 μg size at 200 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Dkk-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.