missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dihydrofolate reductase Protein
A DNA sequence encoding the Homo sapiens (Human) Dihydrofolate reductase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5933-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000782.1 | |
Dihydrofolate reductase Protein | |
Research Use Only | |
DHFR | |
Recombinant Protein | |
E. coli | |
VGSLNCIVAV SQNMGIGKNG DLPWPPLRNE FRYFQRMTTT SSVEGKQNLV IMGKKTWFSI PEKNRPLKGR INLVLSRELK EPPQGAHFLS RSLDDALKLT EQPELANKVD MVWIVGGSSV YKEAMNHPGH LKLFVTRIMQ DFESDTFFPE IDLEKYKLLP EYPGVLSDVQ EEKGIKYKFE VYEKND | |
Greater than 90% as determined by SDS-PAGE. |
1719 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |