missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DHDH Protein
A DNA sequence encoding the Homo sapiens (Human) DHDH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8235-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055290.1 | |
DHDH Protein | |
Research Use Only | |
DHDH | |
Recombinant Protein | |
E. coli | |
MALRWGIVSV GLISSDFTAV LQTLPRSEHQ VVAVAARDLS RAKEFAQKHD IPKAYGSYEE LAKDPSVEVA YIGTQHPQHK AAVMLCLAAG KAVLCEKPTG VNAAEVREMV AEARSRALFL MEAIWTRFFP ASEALRSVLA QGTLGDLRVA RAEFGKNLIH VPRAVDRAQA GGALLDIGIY CVQFTSMVFG GQKPEKISVV GRRHETGVDD TVTVLLQYPG EVHGSFTCSI TVQLSNTASV SGTKGMVQLL NPCWCPTELV VKGEHKEFPL PPVPKDCNFD NGAGMSYEAK HVWECLRKGM KESPVIPLSE SELLADILEE VRKAIGVTFP QDKR | |
Greater than 80% as determined by SDS-PAGE. |
27294 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |