missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Defensin beta 2 Protein

Product Code. 18470922 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18470922 5 μg 5µg
18432822 500 μg 500µg
18434272 1 mg 1mg
18450262 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18470922 Supplier Novus Biologicals™ Supplier No. NBP2349105ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Defensin beta 2 protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using immature human dendritic cells is in a concentration range of 10-100 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15263, O15263, O15263
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 130 mM NaCl.
Gene ID (Entrez) 13215
Molecular Weight (g/mol) 4.3 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of Defensin beta 2 as determined by LAL method.
Gene Alias BD-2, Defb2, Defb2 defensin beta 2, defensin beta 2
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins
Purity or Quality Grade >98% pure by SDS-PAGE and HPLC
Protein Defensin beta 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.