missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DDR2 Protein
A cDNA sequence encoding the DDR2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11611-1mg
Additional Details : Weight : 0.01000kg
Specifications
DDR2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
KAQVNPAICR YPLGMSGGQI PDEDITASSQ WSESTAAKYG RLDSEEGDGA WCPEIPVEPD DLKEFLQIDL HTLHFITLVG TQGRHAGGHG IEFAPMYKIN YSRDGTRWIS WRNRHGKQVL DGNSNPYDIF LKDLEPPIVA RFVRFIPVTD HSMNVCMRVE LYGCVWLDGL VSYNAPAGQQ FVLPGGSIIY LNDSVYDGAV GYSMTEGLGQ LTDGVSGLDD FTQTHEYHVW PGYDYVGWRN ESATNGYIEI MFEFDRIRNF TTMKVHCNNM FAKGVKIFKE VQCYFRSEAS EWEPNAISFP LVLDDVNPSA RFVTVPLHHR MASAIKCQYH FADTWMMFSE ITFQSDAAMY NNSEALPTSP MAPTTYDPML KVDDSNTRLE HHHHHH | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DDR2 solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |