missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human DCPS His Protein

Product Code. 18223102 Shop All Bio Techne Products
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18223102

Brand: Novus Biologicals™ NBP17240650UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-337 of Human DCPS The Recombinant Human DCPS Protein is derived from E. coli. The Recombinant Human DCPS Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96C86
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0) containing 20% glycerol, 1mM DTT
Gene ID (Entrez) 28960
Molecular Weight (g/mol) 40.8kDa
Name DCPS Protein
Quantity 50 μg
Source E. Coli
Immunogen DCPS, 1-337aa, Human, His tag, E.coli (NP_054745)
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.