missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DBNDD1 Protein

A cDNA sequence encoding the DBNDD1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11597-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937758

  • £155.00 / 20µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DBNDD1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMEPPEG AGTGEIVKEA EVPQAALGVP AQGTGDNGHT PVEEEVGGIP VPAPGLLQVT ERRQPLSSVS SLEVHFDLLD LTELTDMSDQ ELAEVFADSD DENLNTESPA GLHPLPRAGY LRSPSWTRTR AEQSHEKQPL GDPERQATVL DTFLTVERPQ ED
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
DBNDD1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers