missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human D-glucuronyl C5-epimerase Protein

Product Code. 15929869
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15929869

Brand: enQuireBio™ QP6087ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) D-glucuronyl C5-epimerase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001311022.1
Gene ID (Entrez) 26035
Name D-glucuronyl C5-epimerase Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol GLCE
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence NKCSSDKAIQ FPRRSSSGFR VDGFEKRAAA SESNNYMNHV AKQQSEEAFP QEQQKAPPVV GGFNSNVGSK VLGLKYEEID CLINDEHTIK GRREGNEVFL PFTWVEKYFD VYGKVVQYDG YDRFEFSHSY SKVYAQRAPY HPDGVFMSFE GYNVEVRDRV KCISGVEGVP LSTQWGPQGY FYPIQIAQYG LSHYSKNLTE KPPHIEVYET AEDRDKNKPN DWTVPKGCFM ANVADKSRFT NVKQFIAPET SEGVSLQLGN TKDFIISFDL KFLTNGSVSV VLETTEKNQL FTIHYVSNAQ LIAFKERDIY YGIGPRTSWS TVTRDLVTDL RKGVGLSNTK AVKPTKIMPK KVVRLIAKGK GFLDNITIST TAHMAAFFAA SDWLVRNQDE KGGWPIMVTR KLGEGFKSLE PGWYSAMAQG QAISTLVRAY LLTKDHIFLN SALRATAPYK FLSEQHGVKA VFMNKHDWYE EYPTTPSSFV LNGFMYSLIG LYDLKETAGE KLGKEARSLY ERGMESLKAM LPLYDTGSGT IYDLRHFMLG IAPNLARWDY HTTHINQLQL LSTIDESPVF KEFVKRWKSY LKGSRAKHN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.