missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human cytosine-5 Protein
A DNA sequence encoding the Homo sapiens (Human) cytosine-5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8293-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001307821.1 | |
cytosine-5 Protein | |
Research Use Only | |
DNMT3A | |
Recombinant Protein | |
E. coli | |
KIMYVGDVRS VTQKHIQEWG PFDLVIGGSP CNDLSIVNPA RKGLYEGTGR LFFEFYRLLH DARPKEGDDR PFFWLFENVV AMGVSDKRDI SRFLESNPVM IDAKEVSAAH RARYFWGNLP GMNRPLASTV NDKLELQECL EHGRIAKFSK VRTITTRSNS IKQGKDQHFP VFMNEKEDIL WCTEMERVFG FPVHYTDVSN MSRLARQRLL GRSWSVPVIR HLF | |
Greater than 90% as determined by SDS-PAGE. |
1788 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |